siRNA Details
Gene Symbol: ZFP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321917.1 | NP_001308846.1 | 5 | 985 | ||
NM_133466.3 | NP_597723.1 | 5 | 982 | |||
GenBank mRNA | AB075828.1 | - | 1 | 2449 | ||
AK074942.1 | - | 5 | 964 | |||
AK315709.1 | - | 5 | 942 | |||
AL834267.1 | - | 5 | 956 | |||
BC032577.2 | - | 5 | 949 | 5492906 | ||
GU388429.1 | - | 5 | 952 |
