siRNA Details
| Gene Symbol: VSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288791.1 | NP_001275720.1 | 4 | 348 | ||
| NM_001288792.1 | NP_001275721.1 | 4 | 557 | |||
| NM_198481.3 | NP_940883.2 | 4 | 557 | |||
| NR_110142.1 | 4 | 557 | ||||
| XM_011526845.1 | XP_011525147.1 | 5 | 864 | |||
| XM_011526846.1 | XP_011525148.1 | 5 | 864 | |||
| XM_011526847.1 | XP_011525149.1 | 5 | 864 | |||
| XM_011526848.1 | XP_011525150.1 | 5 | 864 | |||
| XM_011526849.1 | XP_011525151.1 | 4 | 570 | |||
| XM_017026666.1 | XP_016882155.1 | 4 | 570 | |||
| GenBank mRNA | AY358542.1 | - | 4 | 566 | ||
| BC100943.2 | - | 4 | 534 | 40004095 | ||
| DQ479397.1 | - | 4 | 398 | |||
| FJ584316.1 | - | 4 | 392 | |||
| FJ584317.1 | - | 4 | 392 | |||
| FJ584318.1 | - | 4 | 392 | |||
| FJ882051.1 | - | 4 | 392 | |||
| FN398145.1 | - | 4 | 385 | |||

VSTM1
Genomic Map