siRNA Details
| Gene Symbol: FABP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001442.2 | NP_001433.1 | 2 | 230 | ||
| GenBank mRNA | BC003672.1 | - | 2 | 225 | 3683235 | |
| BQ880795.1 | - | 2 | 162 | |||
| BT006809.1 | - | 2 | 159 | |||
| CD000452.1 | - | 2 | 231 | |||
| CR456903.1 | - | 2 | 159 | |||
| FJ224319.1 | - | 2 | 221 | |||
| J02874.1 | - | 2 | 221 | |||

FABP4
Genomic Map