siRNA Details
| Gene Symbol: FAM19A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004440.1 | NP_001004440.1 | 5 | 824 | ||
| NM_182759.2 | NP_877436.1 | 5 | 756 | |||
| XM_005270766.3 | XP_005270823.1 | 6 | 1043 | |||
| XM_006710582.3 | XP_006710645.1 | 6 | 1111 | |||
| GenBank mRNA | AY325116.1 | - | 5 | 756 | ||
| AY325119.1 | - | 5 | 824 | |||

FAM19A3
Genomic Map