siRNA Details
| Gene Symbol: RIIAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144956.1 | NP_001138428.1 | 1, 2 | 81 | ||
| GenBank mRNA | AA393535.1 | - | 1, 2 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIIAD1| Gene Symbol: RIIAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144956.1 | NP_001138428.1 | 1, 2 | 81 | ||
| GenBank mRNA | AA393535.1 | - | 1, 2 | 71 | ||