siRNA Details
| Gene Symbol: RUSC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039517.1 | NP_001034606.1 | 2 | 632 | ||
| GenBank mRNA | BC062571.1 | - | 2 | 661 | 5183308 | |
| BC080538.1 | - | 2 | 632 | 5018492 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RUSC1-AS1| Gene Symbol: RUSC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039517.1 | NP_001034606.1 | 2 | 632 | ||
| GenBank mRNA | BC062571.1 | - | 2 | 661 | 5183308 | |
| BC080538.1 | - | 2 | 632 | 5018492 | ||