siRNA Details
| Gene Symbol: RSPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038633.3 | NP_001033722.1 | 8 | 2579 | ||
| NM_001242908.1 | NP_001229837.1 | 7 | 2400 | |||
| NM_001242909.1 | NP_001229838.1 | 7 | 2079 | |||
| NM_001242910.1 | NP_001229839.1 | 6 | 2211 | |||
| XM_006710583.3 | XP_006710646.1 | 6 | 2315 | |||
| GenBank mRNA | AK098225.1 | - | 7 | 1823 | ||
| BC114966.1 | - | 6 | 1999 | 40084277 | ||
| DQ165084.1 | - | 7 | 2144 | |||
| DQ165085.1 | - | 6 | 1955 | |||

RSPO1
Genomic Map