siRNA Details
Gene Symbol: LINC00176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027686.1 | 4 | 4641 | |||
NR_027687.1 | 3 | 4611 | ||||
GenBank mRNA | AK130777.1 | - | 2 | 1027 | ||
AK310631.1 | - | 2 | 789 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027686.1 | 4 | 4641 | |||
NR_027687.1 | 3 | 4611 | ||||
GenBank mRNA | AK130777.1 | - | 2 | 1027 | ||
AK310631.1 | - | 2 | 789 |