siRNA Details
| Gene Symbol: SIRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122962.1 | NP_001116434.1 | 3 | 1009 | ||
| NM_001134836.1 | NP_001128308.1 | 3 | 715 | |||
| XM_005260708.4 | XP_005260765.1 | 3 | 818 | |||
| XM_005260709.3 | XP_005260766.1 | 3 | 681 | |||
| XM_011529221.2 | XP_011527523.1 | 3 | 1605 | |||
| XM_011529223.2 | XP_011527525.1 | 4 | 1053 | |||
| XM_011529224.2 | XP_011527526.1 | 3 | 497 | |||
| XM_011529225.2 | XP_011527527.1 | 2 | 418 | |||
| GenBank mRNA | AK095499.1 | - | 3 | 818 | ||
| AK294440.1 | - | 4 | 704 | |||
| AK297072.1 | - | 3 | 609 | |||
| AY424277.1 | - | 3 | 487 | |||

SIRPB2
Genomic Map