siRNA Details
Gene Symbol: GSTTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003082.1 | 3 | 1482 | |||
GenBank mRNA | AK127991.1 | - | 3 | 1482 |
Gene Symbol: GSTTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003081.1 | 3 | 326 | |||
GenBank mRNA | AL050257.1 | - | 3 | 326 |
Gene Symbol: LOC100652871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530666.1 | XP_011528968.1 | 3 | 385 |
