siRNA Details
| Gene Symbol: SEC14L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161368.1 | NP_001154840.1 | 2, 3 | 231 | ||
| NM_174977.3 | NP_777637.1 | 2, 3 | 231 | |||
| XM_006724235.2 | XP_006724298.1 | 2, 3 | 247 | |||
| XM_011530164.2 | XP_011528466.1 | 1, 2 | 362 | |||
| XM_017028787.1 | XP_016884276.1 | 1, 2 | 360 | |||
| XM_017028788.1 | XP_016884277.1 | 1, 2 | 361 | |||
| XM_017028790.1 | XP_016884279.1 | 1, 2 | 368 | |||
| GenBank mRNA | AK092002.1 | - | 2, 3 | 231 | ||
| AK093418.1 | - | 2, 3 | 248 | |||
| AK093448.1 | - | 2, 3 | 215 | |||
| AY158085.1 | - | 2, 3 | 131 | |||
| BC136358.1 | - | 2, 3 | 203 | 9020345 | ||
| BC136359.1 | - | 2, 3 | 203 | 9020346 | ||
| BC139912.1 | - | 2, 3 | 170 | 40147686 | ||
| FJ824055.1 | - | 2, 3 | 148 | |||

SEC14L4
Genomic Map