siRNA Details
| Gene Symbol: CCDC150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080539.1 | NP_001074008.1 | 6 | 883 | ||
| XM_006712438.3 | XP_006712501.1 | 6 | 883 | |||
| XM_011510985.2 | XP_011509287.1 | 5 | 675 | |||
| XM_017003861.1 | XP_016859350.1 | 6 | 883 | |||
| XM_017003862.1 | XP_016859351.1 | 6 | 883 | |||
| XM_017003863.1 | XP_016859352.1 | 6 | 883 | |||
| XM_017003864.1 | XP_016859353.1 | 6 | 883 | |||
| GenBank mRNA | AK096979.1 | - | 6 | 883 | ||

CCDC150
Genomic Map