siRNA Details
| Gene Symbol: FABP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040442.1 | NP_001035532.1 | 4 | 295 | ||
| NM_001130958.1 | NP_001124430.1 | 5 | 375 | |||
| NM_001445.2 | NP_001436.1 | 2 | 210 | |||
| GenBank mRNA | AI208231.1 | - | 4 | 295 | ||
| BC022489.1 | - | 3 | 180 | 4792594 | ||
| DQ132786.1 | - | 5 | 376 | |||
| U19869.1 | - | 2 | 131 | |||
| X90908.1 | - | 2 | 210 | |||

FABP6
Genomic Map