siRNA Details
| Gene Symbol: LOC285095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135766.1 | 2 | 252 | |||
| NR_135767.1 | 2 | 212 | ||||
| NR_135768.1 | 2 | 151 | ||||
| GenBank mRNA | AA250751.1 | - | 2 | 178 | ||
| HY129633.1 | - | 2 | 253 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC285095| Gene Symbol: LOC285095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135766.1 | 2 | 252 | |||
| NR_135767.1 | 2 | 212 | ||||
| NR_135768.1 | 2 | 151 | ||||
| GenBank mRNA | AA250751.1 | - | 2 | 178 | ||
| HY129633.1 | - | 2 | 253 | |||