siRNA Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289947.1 | NP_001276876.1 | 5 | 427 | ||
| NM_001290030.1 | NP_001276959.1 | 10 | 1214 | |||
| XM_011510998.2 | XP_011509300.1 | 12 | 1346 | |||
| XM_011510999.2 | XP_011509301.1 | 10 | 917 | |||
| XM_011511001.2 | XP_011509303.1 | 10 | 1692 | |||
| XM_017003875.1 | XP_016859364.1 | 11 | 1715 | |||
| XM_017003876.1 | XP_016859365.1 | 10 | 730 | |||
| GenBank mRNA | BC127676.1 | - | 5 | 427 | 40127368 | |
| BC150546.1 | - | 5 | 427 | |||
| BQ429225.1 | - | 425 | ||||

ERICH2
Genomic Map