siRNA Details
| Gene Symbol: LINC00636 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015394.1 | 3 | 503 | |||
| GenBank mRNA | BC039517.1 | - | 3 | 503 | ||
| BC101228.2 | - | 3 | 460 | |||
| BC101229.1 | - | 3 | 460 | |||
| BC101230.2 | - | 3 | 460 | |||
| BC101231.1 | - | 3 | 460 | |||

LINC00636
Genomic Map