siRNA Details
| Gene Symbol: RABL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173825.3 | NP_776186.2 | 3, 4 | 307 | ||
| XM_006713595.2 | XP_006713658.1 | 3, 4 | 291 | |||
| XM_017006227.1 | XP_016861716.1 | 3, 4 | 291 | |||
| XM_017006228.1 | XP_016861717.1 | 3, 4 | 291 | |||
| GenBank mRNA | AK293085.1 | - | 3, 4 | 291 | ||
| BC020832.1 | - | 3, 4 | 286 | 4771781 | ||
| BX647559.1 | - | 3, 4 | 304 | |||
| BX647593.1 | - | 3, 4 | 307 | |||
| BX647664.1 | - | 3, 4 | 291 | |||

RABL3
Genomic Map