siRNA Details
| Gene Symbol: FANCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018115.2 | NP_001018125.1 | 19 | 1822 | ||
| NM_001319984.1 | NP_001306913.1 | 19 | 1894 | |||
| NM_033084.4 | NP_149075.2 | 19 | 1822 | |||
| GenBank mRNA | AF230336.1 | - | 19 | 1765 | ||
| AF340183.1 | - | 19 | 1765 | |||
| AK022613.1 | - | 12 | 1224 | |||
| AK307512.1 | - | 20 | 1795 | |||
| AL832427.1 | - | 3 | 201 | |||

FANCD2
Genomic Map