siRNA Details
| Gene Symbol: C3orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007534.2 | NP_001007535.1 | 2 | 1069 | ||
| GenBank mRNA | AK097460.1 | - | 2 | 1069 | ||
| BC137154.1 | - | 2 | 1030 | 9021151 | ||
| BC137155.1 | - | 2 | 1030 | 9021152 | ||

C3orf56
Genomic Map