siRNA Details
| Gene Symbol: ZNF660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173658.2 | NP_775929.2 | 3 | 1049 | ||
| XM_005265090.4 | XP_005265147.1 | 2 | 1147 | |||
| XM_006713117.3 | XP_006713180.1 | 3 | 1148 | |||
| XM_011533622.2 | XP_011531924.1 | 3 | 1352 | |||
| GenBank mRNA | AK094189.1 | - | 3 | 1049 | ||
| BC075094.2 | - | 1 | 740 | 30915605 | ||
| BC075095.2 | - | 1 | 740 | 30915393 | ||
| BX538171.1 | - | 1 | 474 | |||

ZNF660
Genomic Map