siRNA Details
| Gene Symbol: FANCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018115.2 | NP_001018125.1 | 33 | 3401 | ||
| NM_001319984.1 | NP_001306913.1 | 33 | 3473 | |||
| NM_033084.4 | NP_149075.2 | 33 | 3401 | |||
| GenBank mRNA | AF230336.1 | - | 33 | 3344 | ||
| AF340183.1 | - | 33 | 3344 | |||
| AK074406.1 | - | 6 | 599 | |||
| AL832427.1 | - | 17 | 1780 | |||

FANCD2
Genomic Map