siRNA Details
| Gene Symbol: PRRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318871.1 | NP_001305800.1 | 3 | 1288 | ||
| NM_207351.4 | NP_997234.3 | 3 | 1288 | |||
| NR_134900.1 | 3 | 1288 | ||||
| XM_011533628.2 | XP_011531930.1 | 3 | 1366 | |||
| XM_011533629.2 | XP_011531931.1 | 3 | 1346 | |||
| XM_017006256.1 | XP_016861745.1 | 2 | 3153 | |||
| GenBank mRNA | AK090993.1 | - | 3 | 1267 | ||
| AY358158.1 | - | 3 | 1275 | |||
| BC040508.1 | - | 3 | 1265 | 5300209 | ||
| BC089447.1 | - | 3 | 1247 | 30563721 | ||
| BC111555.1 | - | 2 | 1185 | |||

PRRT3
Genomic Map