siRNA Details
| Gene Symbol: DPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047434.2 | NP_001040899.1 | 2 | 412 | ||
| NM_206831.2 | NP_996662.1 | 3 | 487 | |||
| GenBank mRNA | AK022970.1 | - | 2 | 392 | ||
| AK092040.1 | - | 3 | 440 | |||
| AK311618.1 | - | 3 | 449 | |||
| BC010181.2 | - | 3 | 396 | 4543414 | ||
| DA447430.1 | - | 2 | 402 | |||
| DA875039.1 | - | 3 | 487 | |||

DPH3
Genomic Map