siRNA Details
| Gene Symbol: DOK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164673.1 | NP_001158145.1 | 7 | 2504 | ||
| NM_001256896.1 | NP_001243825.1 | 4 | 2213 | |||
| NM_173660.4 | NP_775931.3 | 7 | 2515 | |||
| GenBank mRNA | AK091037.1 | - | 7 | 2499 | ||
| BC141852.1 | - | 7 | 2486 | 8860352 | ||

DOK7
Genomic Map