siRNA Details
| Gene Symbol: DOK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164673.1 | NP_001158145.1 | 7 | 2012 | ||
| NM_001256896.1 | NP_001243825.1 | 4 | 1721 | |||
| NM_173660.4 | NP_775931.3 | 7 | 2023 | |||
| GenBank mRNA | AK091037.1 | - | 7 | 2007 | ||
| AK096456.1 | - | 4 | 1721 | |||
| BC043568.1 | - | 3 | 1360 | |||
| BC062369.1 | - | 3 | 1415 | |||
| BC131544.1 | - | 7 | 1960 | 40085498 | ||
| BC141852.1 | - | 7 | 1994 | 8860352 | ||

DOK7
Genomic Map