siRNA Details
| Gene Symbol: RNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173662.2 | NP_775933.1 | 3 | 589 | ||
| XM_005262938.3 | XP_005262995.1 | 3 | 1326 | |||
| XM_011531879.2 | XP_011530181.1 | 3 | 630 | |||
| XM_011531881.2 | XP_011530183.1 | 3 | 673 | |||
| XM_011531882.2 | XP_011530184.1 | 3 | 1337 | |||
| XM_011531883.2 | XP_011530185.1 | 3 | 650 | |||
| GenBank mRNA | AK091509.1 | - | 3 | 364 | ||
| BC034385.1 | - | 3 | 590 | 5180678 | ||

RNF175
Genomic Map