siRNA Details
Gene Symbol: RNF175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173662.2 | NP_775933.1 | 8 | 1194 | ||
XM_005262939.3 | XP_005262996.1 | 7 | 996 | |||
XM_005262940.4 | XP_005262997.1 | 6 | 1133 | |||
XM_011531879.2 | XP_011530181.1 | 8 | 1139 | |||
XM_011531881.2 | XP_011530183.1 | 8 | 1278 | |||
XM_011531883.2 | XP_011530185.1 | 8 | 1159 | |||
XM_017008047.1 | XP_016863536.1 | 6 | 912 | |||
GenBank mRNA | BC034385.1 | - | 8 | 1195 | 5180678 |
