siRNA Details
| Gene Symbol: RNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173662.2 | NP_775933.1 | 5 | 787 | ||
| XM_005262938.3 | XP_005262995.1 | 5 | 1524 | |||
| XM_005262939.3 | XP_005262996.1 | 4 | 589 | |||
| XM_005262940.4 | XP_005262997.1 | 3 | 726 | |||
| XM_011531879.2 | XP_011530181.1 | 5 | 732 | |||
| XM_011531881.2 | XP_011530183.1 | 5 | 871 | |||
| XM_011531882.2 | XP_011530184.1 | 5 | 1535 | |||
| XM_011531883.2 | XP_011530185.1 | 5 | 752 | |||
| XM_017008047.1 | XP_016863536.1 | 3 | 505 | |||
| GenBank mRNA | AK091509.1 | - | 4 | 407 | ||
| BC034385.1 | - | 5 | 788 | 5180678 | ||

RNF175
Genomic Map