siRNA Details
| Gene Symbol: STPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174952.2 | NP_777612.1 | 9 | 1159 | ||
| XM_011531886.2 | XP_011530188.1 | 11 | 1659 | |||
| XM_017008049.1 | XP_016863538.1 | 9 | 1434 | |||
| XM_017008050.1 | XP_016863539.1 | 9 | 1392 | |||
| XM_017008051.1 | XP_016863540.1 | 6 | 637 | |||
| GenBank mRNA | BC036870.1 | - | 9 | 1159 | 5226859 | |
| BX648305.1 | - | 3 | 211 | |||

STPG2
Genomic Map