siRNA Details
| Gene Symbol: ACSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004457.3 | NP_004448.2 | 8 | 1368 | ||
| NM_203372.1 | NP_976251.1 | 7 | 1261 | |||
| XM_017003584.1 | XP_016859073.1 | 6 | 1180 | |||
| GenBank mRNA | AB061712.1 | - | 8 | 1336 | ||
| AK001471.1 | - | 3 | 287 | |||
| AK023191.1 | - | 5 | 913 | |||
| AK314621.1 | - | 8 | 1346 | |||
| BC041692.1 | - | 8 | 1360 | 5226905 | ||
| D89053.1 | - | 6 | 1059 | |||
More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
Citations (1)
Activation of LXR increases acyl-CoA synthetase activity through direct regulation of ACSL3 in human placental trophoblast cells.
- Authors
- Weedon-Fekjaer MS, Dalen KT, Solaas K, Staff AC, Duttaroy AK, Nebb HI
- Journal
- J Lipid Res - Page No 1886 (Thu Jul 01 00:00:00 UTC 2010)
- Volume (Year)
- 51 (2010)
- Product Usage
- -

ACSL3
Genomic Map