siRNA Details
| Gene Symbol: RNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113561.2 | NP_001107033.1 | 4 | 962 | ||
| NM_001323292.1 | NP_001310221.1 | 4 | 962 | |||
| NM_178532.4 | NP_848627.1 | 4 | 962 | |||
| XM_017009383.1 | XP_016864872.1 | 4 | 962 | |||
| XM_017009384.1 | XP_016864873.1 | 3 | 1196 | |||
| XM_017009385.1 | XP_016864874.1 | 3 | 1195 | |||
| XM_017009386.1 | XP_016864875.1 | 4 | 962 | |||
| XM_017009387.1 | XP_016864876.1 | 4 | 962 | |||
| XM_017009388.1 | XP_016864877.1 | 4 | 921 | |||
| XM_017009389.1 | XP_016864878.1 | 4 | 958 | |||
| GenBank mRNA | AK090756.1 | - | 4 | 961 | ||
| AL832580.2 | - | 4 | 921 | |||
| BC101277.2 | - | 4 | 925 | 40024386 | ||
| BC101278.2 | - | 4 | 925 | 40024390 | ||
| BC101397.2 | - | 4 | 921 | 40024392 | ||

RNF180
Genomic Map