siRNA Details
| Gene Symbol: SREK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173829.3 | NP_776190.1 | 3 | 286 | ||
| XM_011543344.2 | XP_011541646.1 | 2 | 418 | |||
| GenBank mRNA | AK094073.1 | - | 286 | |||
| AL832348.1 | - | 3 | 243 | |||
| BC047006.1 | - | 3 | 215 | |||
| BC108716.1 | - | 3 | 216 | 6184559 | ||
| BC127085.1 | - | 3 | 224 | 40122236 | ||
| BC127086.1 | - | 3 | 224 | 40122239 | ||

SREK1IP1
Genomic Map