siRNA Details
Gene Symbol: 8030451A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020205.1 | - | 14 | |||
AK033210.1 | - | 120 | ||||
AK038610.1 | - | 34 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 8030451A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020205.1 | - | 14 | |||
AK033210.1 | - | 120 | ||||
AK038610.1 | - | 34 |