siRNA Details
| Gene Symbol: CCDC18-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034089.1 | 5 | 447 | |||
| GenBank mRNA | AK021668.1 | - | 1 | 761 | ||
| AL832786.1 | - | 5 | 447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC18-AS1| Gene Symbol: CCDC18-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034089.1 | 5 | 447 | |||
| GenBank mRNA | AK021668.1 | - | 1 | 761 | ||
| AL832786.1 | - | 5 | 447 | |||