siRNA Details
| Gene Symbol: SLC2A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033967.1 | 2 | 590 | |||
| GenBank mRNA | AK056786.1 | - | 2 | 535 | ||
| BU657382.1 | - | 2 | 597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A1-AS1| Gene Symbol: SLC2A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033967.1 | 2 | 590 | |||
| GenBank mRNA | AK056786.1 | - | 2 | 535 | ||
| BU657382.1 | - | 2 | 597 | |||