siRNA Details
| Gene Symbol: LOC100506098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134567.1 | 5 | 625 | |||
| NR_134568.1 | 5 | 534 | ||||
| NR_134569.1 | 4 | 463 | ||||
| GenBank mRNA | BG777800.1 | - | 4 | 464 | ||
| BI759100.1 | - | 5 | 525 | |||
| DR423379.1 | - | 5 | 610 | |||

LOC100506098
Genomic Map