siRNA Details
| Gene Symbol: LOC100996579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110163.1 | 3 | 632 | |||
| NR_110164.1 | 2 | 588 | ||||
| GenBank mRNA | AK027541.1 | - | 2 | 588 | ||
| BG697582.1 | - | 3 | 597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996579| Gene Symbol: LOC100996579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110163.1 | 3 | 632 | |||
| NR_110164.1 | 2 | 588 | ||||
| GenBank mRNA | AK027541.1 | - | 2 | 588 | ||
| BG697582.1 | - | 3 | 597 | |||