siRNA Details
| Gene Symbol: CAGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170692.1 | NP_001164163.1 | 5 | 2142 | ||
| NM_001170693.1 | NP_001164164.1 | 5 | 2142 | |||
| NM_205864.2 | NP_995586.1 | 4 | 1924 | |||
| GenBank mRNA | AF414185.1 | - | 4 | 1923 | ||
| AK097760.1 | - | 5 | 1841 | |||
| BC026188.1 | - | 5 | 1815 | |||
| BC026194.2 | - | 5 | 1827 | 4826784 | ||
| BC033545.1 | - | 5 | 1827 | 4826548 | ||

CAGE1
Genomic Map