siRNA Details
| Gene Symbol: PRR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175922.3 | NP_787118.2 | 1 | 1791 | ||
| GenBank mRNA | BC034775.1 | - | 1 | 1791 | 5177440 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR18| Gene Symbol: PRR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175922.3 | NP_787118.2 | 1 | 1791 | ||
| GenBank mRNA | BC034775.1 | - | 1 | 1791 | 5177440 | |