siRNA Details
| Gene Symbol: LINC00174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026873.1 | 5 | 2927 | |||
| GenBank mRNA | AK091213.1 | - | 1 | 1585 | ||
| AK092625.1 | - | 1 | 704 | |||
| AK124597.1 | - | 1 | 2048 | |||
| BC020399.1 | - | 5 | 2929 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00174| Gene Symbol: LINC00174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026873.1 | 5 | 2927 | |||
| GenBank mRNA | AK091213.1 | - | 1 | 1585 | ||
| AK092625.1 | - | 1 | 704 | |||
| AK124597.1 | - | 1 | 2048 | |||
| BC020399.1 | - | 5 | 2929 | |||