siRNA Details
| Gene Symbol: ZNF775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173680.3 | NP_775951.2 | 3 | 1882 | ||
| GenBank mRNA | BC015152.2 | - | 2 | 1820 | 3546283 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF775| Gene Symbol: ZNF775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173680.3 | NP_775951.2 | 3 | 1882 | ||
| GenBank mRNA | BC015152.2 | - | 2 | 1820 | 3546283 | |