siRNA Details
| Gene Symbol: ATG9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317056.1 | NP_001303985.1 | 4 | 792 | ||
| NR_073169.1 | 5 | 399 | ||||
| NR_133652.1 | 4 | 792 | ||||
| XM_011516065.1 | XP_011514367.1 | 4 | 792 | |||
| XM_011516066.2 | XP_011514368.1 | 4 | 792 | |||
| GenBank mRNA | AK096734.1 | - | 4 | 792 | ||
| AY515311.1 | - | 4 | 792 | |||
| BC128587.1 | - | 5 | 365 | 40129083 | ||

ATG9B
Genomic Map