siRNA Details
| Gene Symbol: FBXO43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029860.3 | NP_001025031.2 | 2 | 2080 | ||
| NR_036491.1 | 2 | 2124 | ||||
| XM_011516987.2 | XP_011515289.1 | 2 | 2026 | |||
| XM_011516989.2 | XP_011515291.1 | 2 | 2070 | |||
| XM_011516990.1 | XP_011515292.1 | 2 | 1296 | |||
| XM_011516992.2 | XP_011515294.1 | 2 | 2026 | |||
| XM_017013358.1 | XP_016868847.1 | 2 | 2026 | |||
| GenBank mRNA | AK310518.1 | - | 2 | 1650 | ||
| BC090892.1 | - | 2 | 2053 | |||

FBXO43
Genomic Map