siRNA Details
| Gene Symbol: CFAP157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012502.2 | NP_001012520.2 | 4 | 704 | ||
| XM_006717064.3 | XP_006717127.2 | 4 | 1432 | |||
| XM_011518559.2 | XP_011516861.2 | 4 | 1435 | |||
| XM_017014629.1 | XP_016870118.1 | 4 | 1432 | |||
| XM_017014630.1 | XP_016870119.1 | 4 | 771 | |||
| GenBank mRNA | AK094948.1 | - | 4 | 628 | ||
| AK126437.1 | - | 2 | 133 | |||
| BC141809.1 | - | 4 | 677 | 40147564 | ||

CFAP157
Genomic Map