siRNA Details
| Gene Symbol: SPATA31E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178828.4 | NP_849150.3 | 4 | 1404 | ||
| GenBank mRNA | AK093206.1 | - | 2 | 872 | ||
| AK125845.1 | - | 4 | 1404 | |||
| AL834438.1 | - | 1 | 502 | |||
| BC137349.1 | - | 4 | 1400 | 9021347 | ||

SPATA31E1
Genomic Map