siRNA Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001022.3 | NP_001013.1 | 2, 3 | 447 | ||
| NM_001321483.1 | NP_001308412.1 | 2, 3 | 124 | |||
| NM_001321484.1 | NP_001308413.1 | 2, 3 | 254 | |||
| NM_001321485.1 | NP_001308414.1 | 2, 3 | 447 | |||
| XM_017027113.1 | XP_016882602.1 | 2, 3 | 442 | |||
| GenBank mRNA | AK311786.1 | - | 2, 3 | 110 | ||
| BC000023.2 | - | 2, 3 | 144 | 3504083 | ||
| BC007615.2 | - | 1, 2 | 75 | 3354280 | ||
| BC017386.1 | - | 2, 3 | 113 | |||
| BC018616.1 | - | 2, 3 | 447 | 4867036 | ||
| BG398397.1 | - | 2, 3 | 254 | |||
| BM468237.1 | - | 2, 3 | 218 | |||
| BP270469.1 | - | 2, 3 | 110 | |||
| BU603942.1 | - | 2, 3 | 113 | |||
| CB124389.1 | - | 2, 3 | 153 | |||
| CB125125.1 | - | 2, 3 | 124 | |||
| M81757.1 | - | 2, 3 | 97 | |||

RPS19
Genomic Map