siRNA Details
Gene Symbol: C9orf142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183241.1 | NP_899064.1 | 6, 7 | 598 | ||
GenBank mRNA | BC002613.2 | - | 6, 7 | 598 | 3161564 | |
BC038191.1 | - | 5, 6 | 749 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C9orf142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183241.1 | NP_899064.1 | 6, 7 | 598 | ||
GenBank mRNA | BC002613.2 | - | 6, 7 | 598 | 3161564 | |
BC038191.1 | - | 5, 6 | 749 |