siRNA Details
| Gene Symbol: GNPTAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024312.4 | NP_077288.2 | 7 | 852 | ||
| XM_006719593.2 | XP_006719656.1 | 7 | 847 | |||
| XM_011538731.2 | XP_011537033.1 | 7 | 848 | |||
| XM_017019961.1 | XP_016875450.1 | 7 | 749 | |||
| XM_017019962.1 | XP_016875451.1 | 7 | 831 | |||
| GenBank mRNA | AK056137.1 | - | 7 | 723 | ||
| AL832172.1 | - | 3 | 294 | |||
| AM085438.1 | - | 7 | 839 | |||
| AY687932.1 | - | 7 | 824 | |||
| BC071687.1 | - | 7 | 800 | |||
| BC131787.1 | - | 7 | 780 | 40126580 | ||

GNPTAB
Genomic Map