siRNA Details
| Gene Symbol: TPRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128228.2 | NP_001121700.2 | 2 | 1738 | ||
| GenBank mRNA | AK074735.1 | - | 2 | 1132 | ||
| BC023530.2 | - | 2 | 746 | |||
| BC048807.1 | - | 2 | 442 | |||
| BC071831.1 | - | 2 | 1068 | |||
| BC098411.1 | - | 2 | 1176 | |||
| BC111500.1 | - | 2 | 867 | 4992533 | ||
| BC128601.1 | - | 2 | 883 | 40132958 | ||
| BC143385.1 | - | 2 | 900 | 9051891 | ||

TPRN
Genomic Map