siRNA Details
| Gene Symbol: TUSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004125.2 | NP_001004125.1 | 1 | 1956 | ||
| GenBank mRNA | AY168647.2 | - | 1 | 1538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TUSC1| Gene Symbol: TUSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004125.2 | NP_001004125.1 | 1 | 1956 | ||
| GenBank mRNA | AY168647.2 | - | 1 | 1538 | ||